Filterpapir, kvantitativt, askefritt
Leverandør: Ahlstrom-Munksjö
100036 100105 100007 100050 100047 100062 100055 100070 100068
512-6026EA 0 NOK
512-6026 512-6029 512-6002 512-6034 512-6037 512-6042 512-6049 512-6173 512-6172
Filterpapir, kvantitativt, askefritt
Filter Filterpapir Kvantitativt filterpapir
Ahlstrom-Munksjö manufactures a series of extremely high purity, acid washed filter papers designed for use in analytical and gravimetric analysis. The quantitative grades are manufactured from top quality cotton linters using ultra-pure, deionised water and are further treated with dilute acid to remove any remaining organic and inorganic impurities. All quantitative grades are manufactured in a strictly controlled environment that ensures high uniformity and high purity from filter to filter.
These filters are treated with a strong acid to remove trace metals and produce high wetMore Product Information