Filterpapir, Polyamidfilter, SUSP.70
Leverandør: Ahlstrom-Munksjö
460001 460002 460003
512-6151EA 1850 NOK
512-6151 512-6150 512-6149
Filterpapir, Polyamidfilter, SUSP.70
Filter Glassfiberfilter
SUSP 70 is a woven polyamide fabric with a 70 μm mesh size used to capture coarse particles such as may be found in waste water.