4,4'-Carbonimidoylbis[N,N-dimethylaniline], Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

51362-25MG
51362-25MGEA 1800 NOK
51362-25MG
4,4'-Carbonimidoylbis[N,N-dimethylaniline], Supelco®
4,4'-Carbonimidoylbis[N,N-dimethylaniline]
Formel: C₁₇H₂₁N₃
CAS-nummer: 492-80-8
EINECS: 207-762-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57769b0f21b7c59528b2c264cbe85b14e8986a91&vid=12a9c2a55679685fcb306f4150d38a52800f1481&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR