Supelco® TraceCERT® Organic Reference Standard, ʟ-Asparagine
Leverandør: Merck
51363-100MG
51363-100MGEA 920 NOK
51363-100MG
Supelco® TraceCERT® Organic Reference Standard, ʟ-Asparagine
Standarder Organiske standarder
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8735376ec12a2886c51a506680fef83349410f7&vid=e8735376ec12a2886c51a506680fef83349410f7&prid=web&clid=SAPDAM