(±)-β-Citronellol, Supelco®

Leverandør: Merck

Synonymer: DL-Citronellol, (±)-Citronellol, Citronellol, β-Citronellol, 3,7-Dimethyloct-6-en-1-ol

51381-1ML
51381-1MLEA 3800 NOK
51381-1ML
(±)-β-Citronellol, Supelco®
(±)-β-Citronellol
Formel: C₁₀H₂₀O
MW: 156.27 g/mol
Kokepunkt: 222
Tetthet: 0.86 g/cm³ (20 °C)
Flash punkt: 100
MDL nummer: MFCD00002935
CAS-nummer: 106-22-9
EINECS: 203-375-0
FN: 3082
ADR: 9 III
Merck Indeks: 13,02354
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a34cc0050055588060f03aad2e4c03f3115f661&vid=9a34cc0050055588060f03aad2e4c03f3115f661&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR