Formaldehyd monitor
Leverandør: 3M
3721
514-0008EA 8250 NOK
514-0008
Formaldehyd monitor
Gassdeteksjonssystemer
Monitors use the proven scientific principle of diffusion, which involves the gradual mixing of molecules of two or more substances.
  • Formaldehyde monitor uses a bisulphite-impregnated filter and is analysed by colorimetry
  • Can be used for eight-hour TWA sampling
  • Does not have the sensitivity for short-term exposure limit (STEL) monitoring
Designed for measuring personal exposure in work areas in a variety of industries.
18-month shelf life.
Sertifiseringer: Meet the OSHA requirement of ±25% accuracy at 0,5 - 5,0 ppm.