In-Line filters, Polydisc™ GW, Whatman™
Leverandør: Whatman products (Cytiva)
10463400 10463401
514-0088EA 5925 NOK
514-0088 514-0089
In-Line filters, Polydisc™ GW, Whatman™
Filter Filter, integrerte
Cytiva's Polydisc GW (Ground Water) is an in-line filter specifically designed for preparation of ground water samples for dissolved heavy metal analysis.
  • Hydrophilic, durable nylon membrane is compatible with aqueous samples
  • 100% quartz fiber prefilter with low heavy metal content allows filtration of larger volumes of even heavily particle-laden samples
  • Certified low background metal values for heavy metal testing
  • Large 20,4 cm² filtration area
  • Tubing nozzle compatible with 6 - 14 mm Int.Ø hose
Polydisc GW’s large filter surface, durable nylonMore Product Information