Dialysis cassettes, Slide-A-Lyzer™ G2, Pierce™
Leverandør: Thermo Fisher Scientific
87731 87728 88251
514-0205EA 2400 NOK
514-0205 514-0202 PIER88251
Dialysis cassettes, Slide-A-Lyzer™ G2, Pierce™
Slanger Dialyseslanger
The Slide-A-Lyzer® G2 dialysis cassette offers efficiency, convenience and sample protection in one package. Sample loading and removal are easily accomplished by using a serological pipette or hypodermic needle attached to a syringe. The built-in air chamber provides sample buoyancy and vertical orientation of the cassette during dialysis.
  • Pipette-accessible for easy sample loading and retrieval
  • Self-floating chambers for buoyancy and vertical orientation during dialysis
  • Designed to maintain the highest possible sample integrity and protection with