Dialysis membrane tubing, Biotech RC
Leverandør: Spectrum Laboratories
133336 133342 133264 133270 133198 133192
514-0639EA 7500 NOK
514-0639 514-0640 514-0636 514-0637 514-0634 514-0633
Dialysis membrane tubing, Biotech RC
Slanger Dialyseslanger
Biotech RC (Regenerated Cellulose) dialysis tubing is crafted from a regeneration process that yields superior temperature tolerances and chemical compatibilities with the same high purity and selectivity as Biotech CE. RC membrane can tolerate concentrated-weak acids & bases, dilute-strong acids and bases, most alcohols and some mild or dilute organic solvents. Exposure to strong polar or organic solvents may damage RC membranes. Use with pH 2 - 12 and temperature 4 - 60 °C.
  • Superior chemical compatibility and temperature tolerance
  • Rigidly controlled