Syringe filters, Minisart® Ophthalsart CE
Leverandør: Sartorius
17528-K
514-0675EA 3725 NOK
514-0675
Syringe filters, Minisart® Ophthalsart CE
Filter Sprøytefilter
Minisart® Ophthalsart with surfactant-free cellulose acetate (SFCA) is made for aqueous or oily solutions with a pH of 4 - 8. Type 17528 with a pore size of 0,2 µm can be used for sterile filtration and has a male luer slip outlet. Version K is individually packed and sterilised by ethylene oxide (EO). Only available in the EU/EEA and registered countries.
  • Sterile and ready-to-use
  • SFCA in many pore sizes
  • Very low non-specific binding
  • PVC-free
  • Bidirectional use possible
Individually packed and sterile Minisart® Ophthalsart syringe filters are optimalMore Product Information