Membrane filters, non sterile, ReliaDisc™
Leverandør: Ahlstrom-Munksjö
760211 760210 760102 760104 760105 760107 760150 760152 760153 760171 760181 760202 760203 760204 760206 760208 760209 760212 760106 760151 760207
516-0797EA 815 NOK
516-0797 514-0732 516-0782 516-0783 516-0784 516-0785 516-0786 516-0787 516-0788 516-0789 516-0790 516-0791 516-0792 516-0793 516-0794 516-0795 516-0796 516-0798 514-0792 514-0793 514-0794
Membrane filters, non sterile, ReliaDisc™
Filter Membranfilter
These plain white non sterile microfiltration membranes are used to remove particles and microorganisms from various organic solvents and aqueous solutions.
  • Particle retention and collection
  • Sterile filtration of solutions
  • Particulate analysis
  • Air monitoring applications
  • Microbiological analysis of potable and waste waters
Cellulose acetate: Low protein-binding, well suited for sterile filtration and clarification of aqueous solutions in the pH range 4 - 8, nutrient media, buffers, and sera. Also compatible with alcohol and oil. These hydrophilicMore Product Information