Pipette controller, Falcon®
Leverandør: Corning
357469
514-1457EA 615 NOK
514-1457
Pipette controller, Falcon®
Pipetter Pipettefyllere
This light, ergonomic, motorised pipette controller is designed for use with glass and plastic serological pipettes from 0,5 to 100 ml.
  • Ergonomic design for exceptional comfort
  • Nosepiece with autoclavable pipette holder and filter
  • Ergonomically positioned finger triggers provide comfortable grip
  • Conveniently positioned switches change operating modes and speeds quickly to handle different liquid volumes and viscosities
  • LCD display for continuous visualisation of speed, modes, and battery status
  • Easy access to the battery compartment
  • Filters, pipette holder