Mixed cellulose ester membranes, non-sterile, 0,2 µm, Whatman™
Leverandør: Whatman products (Cytiva)
10401706 10401712 10401714 10401726 10401731 10401721
514-2161EA 2300 NOK
514-2161 514-2162 514-2163 514-0764 514-0765 514-0945
Mixed cellulose ester membranes, non-sterile, 0,2 µm, Whatman™
Filter Membranfilter
Whatman non-sterile mixed cellulose ester (MCE) filters from Cytiva are suitable for E. coli culturing, enumeration of bacteria, and other culturing and cell counting methods.
  • Plain and gridded options for ease of colony and cell counting methods
  • Supports visual enumeration of bacteria
  • Appropriate for bacterial culturing, including E. coli culturing
  • Biologically inert with good thermal stability
  • Uniform microporous structure results in high flow rates when used for filtering
Whatman MCE filters from Cytiva business are composed of cellulose acetateMore Product Information