Bacterial air venting filters
Leverandør: Cytiva (Formerly Pall Lab)
4210-N 4308-N
514-4107EA 4225 NOK
514-4107 514-4114
Bacterial air venting filters
Filter Ventilasjonsfilter
Economical, disposable depth filter for venting applications. Hydrophobic media allows air and gases to pass freely while blocking aqueous fluid and aerosol contaminents.
  • Use as a vent device for receiving vessels and small isolation or environmental chambers
  • Recommended for small-volume venting and degassing
Hydrophobic glass laminated (polyester/glass fibre/polyester) membrane; PP housing.
Aerosol retention: 99,97% 0,3 µm (DOP) at 32 L/min/100 cm²
* Sterilised by gamma irradiation