Venting filters, Acro® 37 TF
Leverandør: Cytiva (Formerly Pall Lab)
4464 4465
514-4117EA 3000 NOK
514-4117 514-4118
Venting filters, Acro® 37 TF
Filter Ventilasjonsfilter
Multipurpose filter for small volume venting and gas filtration, ideal for small bioreactors and fermenters.
  • Broad chemical compatibility
  • Disposable
Hydrophobic membrane PP reinforcened PTFE; PP housing.