Filtreringskapsel, kull
Leverandør: Cytiva (Formerly Pall Lab)
12011
515-0000EA 2475 NOK
515-0000
Filtreringskapsel, kull
Filter Ventilasjonsfilter
Removes odor, colloids, and organics from liquids using activated carbon.
  • Finely divided carbon provides an active surface area of 65000 m²
  • Ready to use and disposable
Contains 100 g of acid-rinsed, activated charcoal. Inlet/outlet connections are ³/₈" FNPT. Includes optional hose barb adapters to accept ¹/₂" int.Ø tubing.
Applications:
- For laboratory and reagent-grade water and chemical filtration
- Protect laboratory from organic solvent odor