Hydrogen peroksid 30%, ULTREX® II, Ultrapure for trace metal analysis, J.T.Baker®

Leverandør: Avantor
Danger

5155-01EA 3450 NOK
5155-01
Hydrogen peroksid 30%, ULTREX® II, Ultrapure for trace metal analysis, J.T.Baker®
Hydrogen peroksid
Formel: H₂O₂
MW: 34.01 g/mol
Kokepunkt: ∼107 °C (1013 hPa)
Smeltepunkt: < 0 °C
Tetthet: 1.11 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011333
CAS-nummer: 7722-84-1
EINECS: 231-765-0
FN: 2014
ADR: 5.1 II
Merck Indeks: 14,04798

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use 25.0 - 35.0%
Assay (H₂O₂) 25.0 - 35.0 %
Appearance Passes Test
Free Acid (µeq/g) ≤0.5
Chloride (Cl) ≤1 ppm
Nitrate (NO₃) ≤2 ppm
Phosphate (PO₄) ≤2 ppm
ACS - Sulfate (SO₄) ≤5 ppm
Trace Impurities - Aluminum (Al) ≤100 ppt
Trace Impurities - Antimony (Sb) ≤1000 ppt
Trace Impurities - Arsenic (As) ≤1000 ppt
Trace Impurities - Barium (Ba) ≤50 ppt
Trace Impurities - Beryllium (Be) ≤1000 ppt
Trace Impurities - Bismuth (Bi) ≤1000 ppt
Trace Impurities - Boron (B) ≤1000 ppt
Trace Impurities - Cadmium (Cd) ≤50 ppt
Trace Impurities - Calcium (Ca) ≤100 ppt
Trace Impurities - Chromium (Cr) ≤50 ppt
Trace Impurities - Cobalt (Co) ≤50 ppt
Trace Impurities - Copper (Cu) ≤50 ppt
Trace Impurities - Gallium (Ga) ≤1000 ppt
Trace Impurities - Iron (Fe) ≤100 ppt
Trace Impurities - Lead (Pb) ≤100 ppt
Trace Impurities - Lithium (Li) ≤50 ppt
Trace Impurities - Magnesium (Mg) ≤50 ppt
Trace Impurities - Manganese (Mn) ≤50 ppt
Trace Impurities - Mercury (Hg) ≤1000 ppt
Trace Impurities - Molybdenum (Mo) ≤1000 ppt
Trace Impurities - Nickel (Ni) ≤50 ppt
Trace Impurities - Potassium (K) ≤100 ppt
Trace Impurities - Silicon (Si) ≤5000 ppt
Trace Impurities - Silver (Ag) ≤50 ppt
Trace Impurities - Sodium (Na) ≤100 ppt
Trace Impurities - Strontium (Sr) ≤50 ppt
Trace Impurities - Tin (Sn) ≤50 ppt
Trace Impurities - Titanium (Ti) ≤1000 ppt
Trace Impurities - Vanadium (V) ≤1000 ppt
Trace Impurities - Zinc (Zn) ≤50 ppt
Trace Impurities - Zirconium (Zr) ≤1000 ppt
Certificate Provided Reports Actual Lot Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR