VWR® Low Nitrogen Content Filters, Grade 521
Leverandør: VWR Collection
516-0316EA 350 NOK
516-0316
VWR® Low Nitrogen Content Filters, Grade 521
Filter Filterpapir Kvantitativt filterpapir
Low nitrogen content filters for determination of nitrogen content in steel and iron samples dissolved in acid. Filtration of fine precipitates to dissolve according Kjeldahl method.
Bestill nå
SPESIFIKASJONER
Vekt 85 g/m²
Environmentally Preferable
Tykkelse 0,17 mm
Filtreringshastighet 650 s (Herzberg)