Membrane filter dispenser, Sentino®
Leverandør: Cytiva (Formerly Pall Lab)
13184
516-0665EA 27100 NOK
516-0665
Membrane filter dispenser, Sentino®
Membrandispensere
Filter dispensers are ideal for high throughput microbiology testing laboratories looking for more ergonomic, faster ways to handle individual membranes. The Sentino® filter dispenser is designed to aseptically dispense one disc of membrane at the touch of a button.
  • Easy to load without jamming. Guide plates assist loading and keep refill web in alignment during use
  • Soft touch keypad for aseptically dispensing membrane discs
  • Designed to hold 200 membrane discs
  • Compact, portable design with miniaturised refill pack
  • Use plugged into main power source or