Membrane filters, gridded, ReliaDisc™
Leverandør: Ahlstrom-Munksjö
760241 760217 760218 760219 760220 760222 760223 760224 760230 760231 760221 760225 760226 760242 760243 760244 760245 760246 760232 760233 760216 760227 760228 760234 760239 760229
516-0899EA 6675 NOK
516-0899 516-0799 516-0891 516-0892 516-0893 516-0894 516-0895 516-0896 516-0897 516-0898 514-0796 514-0797 514-0798 514-0808 514-0809 514-0810 514-0811 514-0812 514-0802 514-0803 514-0795 514-0799 514-0800 514-0804 514-0807 514-0801
Membrane filters, gridded, ReliaDisc™
Filter Membranfilter
These gridded sterile membranes are specifically designed to save time when performing colony counting and microbiological quality control. The plain white non-sterile microfiltration membranes are used to remove particles and microorganisms from various organic solvents and aqueous solutions.
  • Particle retention and collection
  • Sterile filtration of solutions
  • Particulate analysis
  • Air monitoring applications
  • Gridded membrane filters for easy colony counting
  • Microbiological analysis of potable and waste waters
These cellulose nitrate membrane filters setMore Product Information