Mikrobiologimanifold
Leverandør: Cytiva (Formerly Pall Lab)
4889
516-1038EA 32575 NOK
516-1038
Mikrobiologimanifold
Manifold
This microbiology manifold, with modular design, allows you to optimise testing without sacrificing cleanliness.
  • Can be customised to fit workflow, test area and autoclave
  • Slip-fit adapters enable the use of any Pall microbiology device on the same manifold
  • Right- or left-handed operation
With a 3-place manifold and a coupling device, an additional manifold can be added to make a 6-place manifold. A preventative maintenance schedule can be designed without delays to critical testing.
Adapters are required and need to be ordered separately.