Tangential flow filtreringssystemer og kapsler, Minimate™ EVO
Leverandør: Cytiva (Formerly Pall Lab)
OAPMP220UK OAPMPUNV
516-0205EA 53325 NOK
516-0205 516-1085
Tangential flow filtreringssystemer og kapsler, Minimate™ EVO
Filter Systemer for tangentiell flow
For effektiv konsentrering i laboratorieskala, avsalting og bufferbytteprosesser på opptil 1 L.
  • High product recovery
  • Minimal sample loss
  • Greater efficiency
  • Gentle processing
  • Plug and play processing
  • Scalable operation
Versatile, self-contained capsules include all fittings and tubing to simplify installation into filtration set-ups and will work with any laboratory pump. Dual-pressure gauges allow trans-membrane pressure (TMP) differential calculations.
Capsules feature glass-reinforced polypropylene housing, polypropylene screens, housing sealingMore Product Information