Syringe filters, Sartobind® Protein A
Leverandør: Sartorius
93PRAP06HB-12A
516-1186EA 13100 NOK
516-1186
Syringe filters, Sartobind® Protein A
Filter Sprøytefilter
Sartobind® Protein A syringe filters are the ideal tool for rapid quantification of immunoglobulins (Ig). The large pore size allows ten times faster flow rate than conventional columns and a shorter operation time.
  • Ready-to-use devices
  • Higher throughput per millilitre
  • Enclosed Luer adapters allow connection to a liquid chromatography system