Filterholder, vakuum
Leverandør: Sartorius
16309
516-7017EA 13675 NOK
516-7017
Filterholder, vakuum
Trakter Membranfiltertrakter, trakter
Borsilikatglass, med PTFE-ring, kan autoklaveres.
For particle removal from solvents for analytical determinations and purification of solutions. In combination with solvent resistant, hydrophilic RC-membranes with diameters of 50 or 47 mm, ideal for ultra cleaning and degassing solvents and solvent mixtures for HPLC, GC and AA.
Inhoud van trechter 250 ml, met filtraatfles van 1000 ml.