Ultralab™ Systems
Leverandør: Cytiva (Formerly Pall Lab)
FS007X70
516-8200EA 83700 NOK
516-8200
Ultralab™ Systems
Filter Systemer for tangentiell flow
Ultralab™ systems, composed of Ultrareservoir™ containers and Ultrapump™ II pumps, are designed as a rapid, efficient replacement of dialysis for buffer exchange and desalting of macromolecules.
  • Efficient replacement of dialysis
  • Airtight lid
  • Siphoning action
Systems are specifically designed for use with Ultrasette™ lab tangential flow devices (see 29300-988 series).
Ultrapump™ II has a 0 to 600 rpm rated, ¹/₁₀ hp motor with continuous-duty rated drive, and 0 to 10 dial speed control. Pump size: 16,3×34×13,7 cm (6¹³/₃₂×13¹³/₃₂×5¹³/₃₂"). MaximumMore Product Information