1,10-Fenatrolin monohydrat, Millipore®

Leverandør: Merck Millipore Calbiochem
Danger

Synonymer: 1,10-Phenanthroline hydrate

516705-500
516705-500EA 1875 NOK
516705-500
1,10-Fenatrolin monohydrat, Millipore®
1,10-Fenatrolin monohydrat
Formel: C₁₂H₈N₂·1H₂O
MW: 198,22 g/mol
Smeltepunkt: 93…94 °C
Tetthet: 1,259 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149973
CAS-nummer: 5144-89-8
EINECS: 200-629-2
FN: 2811
ADR: 6.1,III
Merck Indeks: 13,07291
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR