Organic reference standard, Putrescine, analytical standard, Supelco®
Leverandør: Merck
51799-100MG
51799-100MGEA 580 NOK
51799-100MG
Organic reference standard, Putrescine, analytical standard, Supelco®
Standarder Organiske standarder
Putrescine is a naturally occurring polyamine, which is produced via breakdown of ornithine amino acid.
Putrescine may be used as a standard in determining the concentration of putrescine present in fish and fishery products using thin layer chromatography (TLC) coupled with densitometry and high performance liquid chromatography (HPLC).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a220a2c3ff8fdd8f406832821e188501dbf4762e&vid=a220a2c3ff8fdd8f406832821e188501dbf4762e&prid=web&clid=SAPDAM