Supelco® Organic Reference Standard, Tri-o-tolyl phosphate

Leverandør: Merck

51885-250MG
51885-250MGEA 675 NOK
51885-250MG
Supelco® Organic Reference Standard, Tri-o-tolyl phosphate
Standarder Organiske standarder

  • GHS signal: GHS08, GHS07, GHS09


NB: P260 - P264 - P270 - P273 - P301 + P312 - P308 + P311

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9b708fbc519e207f5eef555efa5ef68b3a332ca&vid=e9b708fbc519e207f5eef555efa5ef68b3a332ca&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) NSC 438; Phosphoric acid tris(2-methylphenyl) ester; TOTP; Tri-o-cresyl phosphate
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 78-30-8
MDL No. MFCD00041906
Molekylvekt 368,36
Hardhetsklasse beskrivelse H302 - H370 - H411
Flammepunkt 110 °C
Fraktforhold STD
Lagringsforhold 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR