VWR® CompactStar CS4, Sentrifuge
Leverandør: VWR Collection
521-2853EA 14375 NOK
521-2853 521-2854 521-2855
VWR® CompactStar CS4, Sentrifuge
Sentrifuger Sentrifuger, bordmodell
CompactStar CS4-sentrifugen brukes til å separere ulike prøver innenfor kjemi, livsvitenskap, klinisk forskning og rutinemessig diagnose.
  • High speed: Up to 6500 min⁻¹ / 4000×g
  • Control panel featuring easy turn knobs and large digital displays for time and speed
  • Timer can be set up to 30 minutes and to the ‘On’ position for a continuous run
  • The compact footprint makes it ideal for almost any laboratory, and it can easily be transported from lab to lab
The combination of tube adapters are suitable for running 5 ml (12×75 mm) and 7&More Product Information