VWR® CompactStar CS8, Centrifuge
Leverandør: VWR Collection
521-3345EA 11800 NOK
521-3345
VWR® CompactStar CS8, Centrifuge
Sentrifuger Sentrifuger, bordmodell
The CompactStar CS8 is a small ventilated bench top centrifuge designed for the separation of various samples in the fields of chemistry, life science, clinical research and routine diagnostics. The supplied rotor accepts 8×15 ml conical tubes, 8×15 ml (16×125 mm) round bottom and 8×10 ml (16×100 mm) blood collection tubes directly and smaller tubes with an included adapter set.
  • Easy operation with key pad, large digital displays for time and speed (RPM or RCF)
  • Indicator shows if lid is open/closed
  • Quick spin button for short cycle mode
  • Adjustable