VWR® Centrifuge II, Microplate, PCR Plate Centrifuge
Leverandør: VWR Collection
521-3497EA 9950 NOK
521-3497
VWR® Centrifuge II, Microplate, PCR Plate Centrifuge
Sentrifuger Mikrosentrifuger
The PCR Plate Centrifuge II is a personal sized digital centrifuge designed to spin down droplets in PCR plates. Recommended before and after the use of PCR plates in a thermal cycler for optimised concentration and improved PCR yield. This centrifuge is equipped with a dual-position rotor that accepts a wide variety of PCR plates; 96-well, 384-well, skirted and non skirted. Plates are inserted vertically and quickly spin to a max. speed of 2800 rpm (550 ×g).
  • Digital display for speed and time
  • Quick spin function
  • Safety lid lock
  • Electronic braking system