Centrifuges, General Purpose Pro CTS Series
Leverandør: Thermo Scientific
75009054
521-3510EA 0 NOK
521-3510
Centrifuges, General Purpose Pro CTS Series
Sentrifuger Sentrifuger, gulvmodell
General Purpose Pro Centrifuges Cell Therapy Systems (CTS™) Series consist of equipment, documentation, and certifications, as well as compliance services that support Good Manufacturing Practice (GMP) requirements. The CTS Series helps you get up and running faster, stay compliant and current with all necessary regulatory certifications, navigate regulatory audits, and stay on schedule, taking cell therapy from discovery to clinical research and commercial manufacturing. The refrigerated 4 L (4×1000 ml) floor standing and bench models are designed to meet the needs of today’s rapid-fireMore Product Information
Bestill nå
SPESIFIKASJONER
Modell Multifuge X4F R Pro Cell Therapy Package CTS Series
Environmentally Preferable
Type Centrifuge Package
Maks. kapasitet - vinkelrotor 6×250 ml (F14-6×250LE rotor)
Maks. kapasitet - swing-out-rotor 4×1000 ml (TX-1000 rotor)
Maks. RCF - vinkelrotor 25830 ×g (Microlitre 30×2 rotor)
Maks. RCF - swing-out-rotor 7164 ×g with BIOShield 1000A rotor
Maks. hastighet - vinkelrotor 15200 min⁻¹ (F21-48×2, Microlitre 48×2 or Microlitre 30×2)
Maks. hastighet - swing-out-rotor 6000 min⁻¹ (BIOShield 1000A rotor)
Akselerasjons-/bremseprofiler 9/10
Programmer <100
Sound level 52 dB(A)
Ur 99 h, 59 min, 59 s; with continuous and customisable pulse
Temperatur –10...+40 °C
Display Full colour touch screen interface
W×D×H 566×690×835 mm
Weight 145 kg