Supelco® TraceCERT® Organic Reference Standard, trans-Ferulic acid
Leverandør: Merck
52229-50MG
52229-50MGEA 920 NOK
52229-50MG
Supelco® TraceCERT® Organic Reference Standard, trans-Ferulic acid
Standarder Organiske standarder
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
NB: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02b851787c3a4d4753b1aad0af7ee3e4128f024c&vid=02b851787c3a4d4753b1aad0af7ee3e4128f024c&prid=web&clid=SAPDAM