Cetyl trimethylammonium hydrogen sulphate ≥99.0% (by titrimetric analysis) for ioneparkromatografi, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

52371-5G-F
52371-5G-FEA 1690 NOK
52371-5G-F
Cetyl trimethylammonium hydrogen sulphate ≥99.0% (by titrimetric analysis) for ioneparkromatografi, Supelco®
Cetyl trimethylammonium hydrogen sulphate
Formel: C₁₉H₄₃NO₄S
MDL nummer: MFCD00134393
CAS-nummer: 68214-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9804c99fd545a9ca2d3e4a4dafe6aa0f199ab1c&vid=c22964ce66787408f4474544e9df3769746f8210&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR