Supelco® Organic Reference Standard, Anthrone

Leverandør: Merck

52445-100MG
52445-100MGEA 580 NOK
52445-100MG
Supelco® Organic Reference Standard, Anthrone
Standarder Organiske standarder

NB: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f4a716e545d1c5cb8d986d282592d38455715b8&vid=2f4a716e545d1c5cb8d986d282592d38455715b8&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) 9(10H)-Anthracenone
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 90-44-8
MDL No. MFCD00001187
Molekylvekt 194,23
Smeltepunkt 154 - 157 °C (lit.)
Hardhetsklasse beskrivelse H315 - H319 - H335
Fraktforhold STD
Lagringsforhold 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR