VWR® SuperClearUltra-High Performance, Sentrifugerør
Leverandør: VWR Collection
525-0155EA 2800 NOK
525-0155
VWR® SuperClearUltra-High Performance, Sentrifugerør
Rør Sentrifugerør
These popular, conical-bottom centrifuge tubes are designed for high speed centrifugation, and are made of non cytotoxic, medical grade resin that allows users to easily see sample volume and colour.
  • Prevents breakage and leakage during high speed centrifugation
  • Easy to read graduations for accurate measurements
  • Patented extra large, smear resistant writing panel
  • Versatile packaging options
  • Free from RNase, DNase, endotoxin and ATP
SuperClear tubes feature highly visible printed graduations for easy filling and sample preparation. The patented, extraMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact,Sustainable Packaging
Miljøsertifikat