Sentrifugerør
Leverandør: Greiner Bio-One
188280_1000
525-0394EA 6925 NOK
525-0394
Sentrifugerør
Rør Sentrifugerør
PP, brown, with blue HDPE screw cap, sterile (SAL 10⁻³), graduated, conical bottom, autoclavable without cap.
  • Blue graduations with white marking area
  • PP tubes brown coloured for protection of light-sensitive samples
  • DNase, RNase and human DNA free, non-pyrogenic, non-cytotoxic
Temperature range: −196 to +121 °C (−50 to +100 °C for the cap).
RCF max. (angle rotor/ swing-out rotor): 15000×g/4000×g for 15 ml tubes; 9500×g/3200×g for 50 ml tubes.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat ISO 14001 (241002); ISO 50001 (241002)