Sentrifugeflasker, PC/PPCO, superhastighet, med forseglingslokk, Nalgene®
Leverandør: Thermo Fisher Scientific
3140-1006 3141-1006
525-0466EA 0 NOK
525-0466 525-0467
Sentrifugeflasker, PC/PPCO, superhastighet, med forseglingslokk, Nalgene®
Flasker Sentrifugeflasker
Super-speed centrifuge bottles are designed to boost productivity and pellet recovery in super speed large volume gross separation applications.
  • Process a full liter in each bottle at speeds up to 15810×g
  • Smooth inner walls reduce sheering in mammalian culture separations
  • PC (for clarity and mechanical strength) and PP (for chemical resistance)
  • Recommended operating temperature range 4 to 22 °C
  • Storage temperature: –70 °C
Ideal for bacterial, yeast and tissue isolations; improved viral harvesting in vaccine production; reduced shearing in mammalianMore Product Information