Centrifuge tubes, conical, Nunc™
Leverandør: Thermo Fisher Scientific
339650 339651 339653 339652 376814
525-0492EA 4250 NOK
525-0492 525-0493 525-0495 525-0494 525-0915
Centrifuge tubes, conical, Nunc™
Rør Sentrifugerør
PP, conical bottom, graduated, sterile, autoclavable with HDPE screw cap.
  • Leakproof to protect samples
  • USP Class VI, non pyrogenic, non cytotoxic and RNase-/DNase-free
  • Recyclable plastic rack reduces waste and minimises risk of contamination and particles
It provides open, easy access for pipetting, decanting, and removal of the pellet following centrifugation.
Sertifiseringer: USP Class VI.
Forpakning: Tubes are packaged 40 per case.