VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Leverandør: VWR Collection
525-0633EA 2325 NOK
525-0633 525-0635 525-0634
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Rør Sentrifugerør
Manufactured of high-quality USP Class 6 gamma resistent clear polypropylene with HDPE screw cap.
  • Longer length screw caps
  • Easy-to-read black graduations are accurate to ±2%
  • Large unerasable frosting white printed writing area
  • Both the graduations and writing areas are chloroform-resistant
  • Autoclavable at 121 °C and freezable to –80 °C
  • Leak-Proof
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
These disposable freestanding tubes feature black printedMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat ISO 14001/Certificate No.: 04923E00243R0M