VWR®, Microcentrifuge Tubes and Screw Caps
Leverandør: VWR Collection
525-0647EA 955 NOK
525-0647 525-0649 525-0651 525-0643 525-0653 525-0654 525-0655 525-0656 525-0657 525-0658 525-0645 525-0659 525-0641
VWR®, Microcentrifuge Tubes and Screw Caps
Rør Mikrosentrifugerør
Made from high grade gamma resistance PP.
  • Autoclavable at 121 °C and freezable to –80 °C
  • Tubes can be centrifuged to 20000×g
  • DNase/RNase free and non pyrogenic
  • Caps accept colour coded inserts, and easily write on the frosted side of insert
  • O-rings made from silicone to insure a leakproof seal
  • 1,5 and 2,0 ml tubes provide easy and legible frosted area
  • Sustainable design based on virgin resins
Manufactured in a Class 100000 room environment.
Sertifiseringer: ISO 13485:2003 and ISO 9001:2008.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Miljøsertifikat ISO 14001/Certificate No.: 04923E00243R0M ISO 14001/Certificate No.: 04923E00243R0M ISO 14001/Certificate No.: 04923E00243R0M ISO 14001/Certificate No.: 04923E00243R0M ISO 14001/Certificate No.: 04923E00243R0M ISO 14001/Certificate No.: 04923E00243R0M ISO 14001/Certificate No.: 04923E00243R0M ISO 14001/Certificate No.: 04923E00243R0M ISO 14001/Certificate No.: 04923E00243R0M ISO 14001/Certificate No.: 04923E00243R0M ISO 14001/Certificate No.: 04923E00243R0M ISO 14001/Certificate No.: 04923E00243R0M ISO 14001/Certificate No.: 04923E00243R0M