VWR® PolarSafe, Dispenser
Leverandør: VWR Collection
525-0672EA 1500 NOK
525-0672
VWR® PolarSafe, Dispenser
Etikett- og tapedispensere
Addition to the PolarSafe family of products, the label dispenser can accommodate up to six rolls of cryogenic dots or strip labels. Made from durable, easy-to-clean, acrylic plastic, the dispenser has been designed to conveniently dispense labels for your cryogenic vials, tubes or plates.
  • Four rubber feet hold item in place when used on countertops
  • Can also be wall mounted (screws not included)