VWR®, Cryoboxes, PP
Leverandør: VWR Collection
525-0973EA 795 NOK
525-0973
VWR®, Cryoboxes, PP
Bokser Kryobokser
PP, with translucent lid.
  • Temperature resistant to −86 °C
  • Alphanumeric grid for easy sample management
  • Fits standard 76 mm ULT freezer racks
Designed for use with 5 ml conical centrifuge tubes.