VWR®, Microcentrifuge Tubes, Low Retention
Leverandør: VWR Collection
525-1006EA 555 NOK
525-1006
VWR®, Microcentrifuge Tubes, Low Retention
Rør Mikrosentrifugerør
PP, translucent.
  • Leakproof snap caps
  • Non sterile, non sticking
  • Graduated markings
  • Free from RNase and DNase
For use with rDNA and viscous proteins, these low retention graduated polypropylene tubes may be used between –90 and 140 °C.
They feature flat caps and frosted areas on the sides for easy labeling. Leakproof snap caps may be penetrated by a syringe. Easy-to-open caps will not pop open during autoclaving.
The 0,5 ml tubes fit many thermal cyclers.