VWR®, Microcentrifuge Tubes with Flat Screw Caps
Leverandør: VWR Collection
525-1137EA 545 NOK
525-1137 525-1138 525-1139 525-1140 525-1141 525-1143 525-1144 525-1145 525-1146 525-1147 525-1148 525-1149 525-1155 525-1156 525-1157 525-1158 525-1159 525-1160
VWR®, Microcentrifuge Tubes with Flat Screw Caps
Rør Mikrosentrifugerør
These tubes are made from ultra-clear USP Class VI PP that allows easy viewing of contents.
  • RNase-, DNase- and endotoxin-free
  • Engraved graduations ensure accuracy
  • Fit most common rotors
  • Smooth surface for applying labels
  • Sterile tubes are sterilised by irradiation
These tubes feature universal screw threads for use with most popular brand screw caps. Low profile caps include a silicone rubber O-ring, ensuring a leakproof seal. (Please NOTE: Silicone O-rings have been used from June 22, 2022, first lot number: 220622626D. Manufacturer stopped using theMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat ISO 14001/Certificate No.: 04923E00243R0M