VWR®, Microcentrifuge Tubes with Socket Screw Caps
Leverandør: VWR Collection
525-0652EA 1150 NOK
525-0652 525-0646 525-0648 525-0644 525-0650 525-0642 525-1231 525-1233 525-1234 525-1235 525-1236 525-1237 525-1238 525-1239 525-1240 525-1241 525-1243 525-1244
VWR®, Microcentrifuge Tubes with Socket Screw Caps
Rør Mikrosentrifugerør
These clear, high grade PP tubes are ideal for sample processing applications.
  • RNase-, DNase- and endotoxin-free
  • Sterile tubes are sterilised by irradiation
Excellent for freezer storage or secure sample handling. Screw caps incorporate silicone O-rings for a leakproof seal and accept colour coded inserts. Inserts snap into the top of the cap to create a flush surface. Tubes are available with standard or loop caps, which tether the cap to the tube.
Caps and tubes are steam autoclavable at 121 °C and freezable to −80 °C.
Sertifiseringer: More Product Information