VWR®, Multi Tube Racks, Blue/Natural
Leverandør: VWR Collection
525-1591EA 620 NOK
525-1591 525-1592
VWR®, Multi Tube Racks, Blue/Natural
Stativer for rør Tube Racks
These flexible tube racks are designed for use with a multitude of tube diameters and heights. Each rack holds virtually any brand tube between 10 and 30 mm.
  • Rotate tube rack insert to an angle position to facilitate more comfortable pipetting
  • Rack wells have a unique teardrop design that enables better retention of tubes over traditional designs
  • Open design allows for better filling visibility of tube graduations when filling
  • Inserts can be pivoted in either an upper or lower position to securely accommodate the size and height of tube being used