VWR®, Test Tube Racks, Mega, for 18 to 20 mm Tubes
Leverandør: VWR Collection
525-1594EA 1000 NOK
525-1594 525-1595
VWR®, Test Tube Racks, Mega, for 18 to 20 mm Tubes
Stativer for rør Tube Racks
These fully-assembled, polypropylene Mega Racks are great for high throughput labs requiring large sample storage.
  • Stores a high quantity of samples eliminating the need for multiple racks
  • Alphanumeric grid for quick location and retrieval of tubes
Made of sturdy, lightweight material for easy transport.