n-Heksan, BAKER ANALYZED® for analyse av pesticidrester, J.T.Baker®

Leverandør: Avantor

5274.2500EA 2975 NOK
5274.2500
n-Heksan, BAKER ANALYZED® for analyse av pesticidrester, J.T.Baker®
n-Heksan
Formel: H₃C(CH₂)₄CH₃
MDL nummer: MFCD00009520
CAS-nummer: 110-54-3
FN: 1208
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (by GC) min. 95.0 %
Acidity max. 0.0005 meq/g
Evaporation Residue max. 5 ppm
GC sensitive impurities (lindane GC-ECD) max. 5 ng/l
Water (KF) max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR