Aceton, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®

Leverandør: Avantor

Synonymer: 2-Propanone, DMK, Propanone, Dimethyl ketone

5276.2500EA 3075 NOK
5276.2500
Aceton, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Aceton
Formel: CH₃COCH₃
MDL nummer: MFCD00008765
CAS-nummer: 67-64-1
FN: 1090
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (by GC) min. 99.8 %
Acidity max. 0.0005 meq/g
Evaporation Residue max. 5 ppm
GC sensitive impurities (lindane GC-ECD) max. 5 ng/l
Water (KF) max. 0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR